Learn More
Epredia™ HistoScreen™ Cassettes
Shop All Epredia Products
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Pack of 1000
Description
- Surface finish and reinforced print faces are engineered to optimize print quality. Tube design is precisely engineered to reduce jamming and prevent downtime.
- Grossing, Processing and Embedding – cassette design and flow characteristics tested to ensure efficient reagent exchange. Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples.
- Precision engineering for solid connection to microtomes.
- Cassette surface texture optimizes scanability for archiving.
Specifications
Specifications
| Pore Style | Mesh |
| Closure Included | Separate lids included |
| Quantity | 1000 |
| Closure Type | Back Hinge |
| Color | Lavender |
| Product Type | HistoScreen™ Cassette |