Learn More
Epredia™ HistoScreen™ Cassettes
Product Code. 16343926 Shop All Epredia Products
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Pack of 1000
Product Code. 16343926
Brand: Epredia™ B851000GN
For very small biopsy tissue samples with a back-edge lid hinge. Eliminates the need for sponges or biopsy bags.
  • Surface finish and reinforced print faces are engineered to optimize print quality. Tube design is precisely engineered to reduce jamming and prevent downtime.
  • Grossing, Processing and Embedding – cassette design and flow characteristics tested to ensure efficient reagent exchange. Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples.
  • Precision engineering for solid connection to microtomes.
  • Cassette surface texture optimizes scanability for archiving.
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Mesh
Closure Included Separate lids included
Quantity 1000/Cs.
Closure Type Back Hinge
Color Green
Product Type HistoScreen™ Cassette