Learn More
Epredia™ Histoplast Paraffin
Shop All Epredia Products
Click to view available options
Product Type:
Paraffin IM
Paraffin LP
Paraffin PE
Unit Size:
Pack of 8
Description
- Made of paraffin and plastic polymers
- Delivers excellent processing and yields thin, wrinkle-free sections
Packaged in convenient, easy-to-open, resealable 2 lb. bags. Also available in a premeasured paraffin kit for use on the Epredia™ Excelsior™ AS Tissue Processor (waste drawer included).
Specifications
Specifications
| Melting Point | 56°C |
| Product Type | Paraffin IM |