missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: MTENSTDHPK YSDMIVAAIQ AEKNRAGSSR QSIQKYIKSH YKVGENADSQ IKLSIKRLVT TGVLKQTKGV GASGSFRLAK SDEPKKSVAF KKTKKEIKKV ATPKKASK Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 39%.
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a replication-independent histone that is a member of the histone H1 family.
Specifications
Specifications
| Antigen | Histone 1F0 |
| Applications | ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | H1-0 |
| Gene Accession No. | P07305 |
| Gene Alias | D130017D06Rik; H1 histone family member 0; H1 histone family, member 0; H1(0); H1.0, H1(0), H1-0; H10; H1-0 histone; H1f0; H1fv; histone H1'; Histone H1(0); histone H1.0; Histone H1.0, N-terminally processed; Histone H1-0; MyD196 |
| Gene Symbols | H1-0, H1F0 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?