missing translation for 'onlineSavingsMsg'
Learn More

Histamine diphosphate, MP Biomedicals™

Product Code. 11493020 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11493020 5 g 5g
11483020 1 g 1g
11403030 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 11493020

Brand: MP Biomedicals 0210034305

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Kemiske identifikatorer

CAS 51-74-1
Molecular Formula C5H15N3O8P2
Molecular Weight (g/mol) 307.14
MDL Number MFCD00064316
InChI Key ZHIBQGJKHVBLJJ-UHFFFAOYSA-N
Synonym histamine acid phosphate, 2-1h-imidazol-4-yl ethanamine bis phosphate, histamine biphosphate, histamine dihydrogen phosphate, histamine phosphate 1:2, histamine phosphate, 1h-imidazole-4-ethanamine, phosphate 1:2, histamine phosphate usp, histamine positive, 4-2-aminoethyl imidazole bis dihydrogen phosphate
PubChem CID 65513
ChEBI CHEBI:51193
IUPAC Name 2-(1H-imidazol-5-yl)ethan-1-amine; bis(phosphoric acid)
SMILES OP(O)(O)=O.OP(O)(O)=O.NCCC1=CN=CN1

Tekniske data

Color White
Quantity 5 g
Formula Weight 325.15
Physical Form Crystalline Powder
Chemical Name or Material Histamine diphosphate
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.