Learn More
Honeywell™ HIKE (i)XTREM S3 Shoes
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
49
Unit Size:
1 pair
Description
- Water resistant grain full leather
- Ergonomic collar and padded tongue for an optimized comfort
- Gusset to avoid water and dust penetration
- Breathable 3D lining
- Overcap
- Toe cap metal free
- Metal free antiperforation insert
- Injected PU / Nitrile sole
- (i)NTELLIGENT wear control
- EN ISO 20345:2011 S3 HRO HI CI SRC
Specifications
Specifications
| Certifications/Compliance | EN ISO 20345:2011 S3 HRO HI CI SRC |
| Color | Brown, Black |
| Material | Leather |
| Size | 49 |
| Sole Material | Polyurethane and Nitrile |
| Toe Style | Metal-free |