Learn More
Macherey-Nagel™ High Temperature Septa
Brand: Macherey-Nagel™ 702622
Additional Details : Weight : 0.01000kg
Specifications
Red | |
Red Non-bleeding Silicone | |
320°C | |
3 mm |
12 mm | |
High Temperature Septa | |
50 Pk. |