missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ High Temperature Septa

Brand:  Macherey-Nagel™ 702620

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10371532

  • 68.20 EUR / Pack of 50
Estimated Shipment: 08-08-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Red
Red Non-bleeding Silicone
320°C
3 mm
10 mm
High Temperature Septa
50 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers