Learn More
UVP High-Performance Uv Variable Intensity Transilluminator
Product Code. 12803938
Click to view available options
Wavelength:
254 nm
Unit Size:
Each
Product Code. 12803938
Brand: UVP 95042702
Only null left
Add to basket
Add to basket
UVP's high performance transilluminators feature 25W tubes and a specially constructed filter that provides the most uniform light emission possible from a tube based unit.
The units have a stainless steel top that is easy to clean and a hinged UV blocking lid to protect the user. High, medium and low intensity settings make the unit suitable for both visualisation and band cutting. An internal fan improves the tube lifetime.Dimensions: 337mm x 486mm x 143mm
Specifications
Wavelength 254 nm