Learn More
Scotch-Brite™ High Performance Microfibre Wipes 2010
Product Code. 17207630
Click to view available options
Color:
Blue
Green
Red
White
Yellow
Unit Size:
Each
Pack of 50
Product Code. 17207630
Brand: Scotch-Brite™ 7000103437
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
High performance microfibre cloth that can be used on all surfaces, even delicate ones.
  • High performance microfibre wipe offering highly efficient, and lint free cleaning on virtually any surface.
  • High quality cleaning results even when used with or without water, and with or without detergent
  • Extremely durable, and can resist up to 500 wash cycles
  • Machine washable up to 95°C
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 360 mm
Width (Metric) 320 mm
Color Yellow
Dimensions (L x W) 320 x 360 mm
Packaging Quantity 10/Pk.