missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIGD1B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 EUR - 410.00 EUR
Specifications
| Antigen | HIGD1B |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16823955
|
Proteintech
25367-1-AP-20UL |
20 μL |
145.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16813955
|
Proteintech
25367-1-AP-150UL |
150 μL |
410.00 EUR
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| HIGD1B | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9P298 | |
| 51751 | |
| HIGD1B Fusion Protein Ag21871 | |
| Primary | |
| -20°C | |
| HIGD1B |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| CLST11240, CLST11240 15, HIG1 domain family member 1B, HIGD1B, Protein CLST 11240 | |
| HIGD1B | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title