missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIF3A Monoclonal Antibody (OTI4C2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800735
This item is not returnable.
View return policy
Description
Cell growth and viability is compromised by oxygen deprivation (hypoxia). Hypoxia-inducible factors, including HIF-1 alpha, HIF-1 beta (also designated Arnt 1), EPAS-1 (also designated HIF-2 alpha) and HIF-3 alpha, induce glycolysis, erythropoiesis and angiogenesis in order to restore oxygen homeostasis. Hypoxia-inducible factors are members of the Per-Arnt-Sim (PAS) domain transcription factor family. In response to hypoxia, HIF-1 alpha is upregulated and forms a heterodimer with Arnt 1 to form the HIF-1 complex. The HIF-1 complex recognizes and binds to the hypoxia responsive element (HRE) of hypoxia-inducible genes, thereby activating transcription.Specifications
| HIF3A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y2N7 | |
| HIF3A | |
| Human recombit protein fragment corresponding to amino acids 355-669 of human HIF3A produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4C2 | |
| Unconjugated | |
| HIF3A | |
| HIF-3A, HIF3-alpha-1, IPAS, MOP7, PASD7, bHLHe17 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 64344 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction