missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIF3A Monoclonal Antibody (OTI4C2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800735S
This item is not returnable.
View return policy
Description
HIF3A Monoclonal antibody specifically detects HIF3A in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| HIF3A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P06729 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 914 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4C2 | |
| Unconjugated | |
| CD2 | |
| Basic-helix-loop-helix-PAS protein MOP7; BHLHE17; Class E basic helix-loop-helix protein 17; HIF-3 alpha; HIF3 alpha 1; HIF3A; HIF-3A; HIF-3-alpha; HIF3-alpha; HIF3-alpha-1; hypoxia inducible factor 3 alpha; hypoxia inducible factor 3 alpha subunit; hypoxia inducible factor 3, alpha subunit; hypoxia inducible factor 3a; hypoxia inducible factor three alpha; hypoxia-inducible factor 3 alpha; hypoxia-inducible factor 3-alpha; Inhibitory PAS domain protein; IPAS; Member of PAS protein 7; Mop7; Neonatal and embryonic PAS protein; NEPAS; PAS domain-containing protein 7; PASD7 | |
| Full length human recombinant protein of human CD2 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction