missing translation for 'onlineSavingsMsg'
Learn More

HG6-64-1, MedChemExpress

Product Code. 16451307
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16451307 50 mg 50mg
16497787 5 mg 5mg
16455806 100 mg 100mg
16485068 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16451307 Supplier Medchem Express Supplier No. HY12291

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress HG6-64-1 is a potent and selective B-Raf inhibitor extracted from patent WO 2011090738 A2, example 9 (XI-1); has a IC50 of 0.09 μM on B-raf V600E transformed Ba/F3 cells.

HG6-64-1 is a potent and selective B-Raf inhibitor extracted from patent WO 2011090738 A2, example 9 (XI-1)

Specifications

Chemical Name or Material HG6-64-1
Physical Form Solid
CAS 1315329-43-1
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C32H34F3N5O2
For Use With (Application) Cancer-Kinase/protease
Synonym HMSL 10017-101-1
Solubility Information DMSO : ≥ 100 mg/mL (173.12 mM)
SMILES O=C(NC1=CC=C(CN2CCN(CC)CC2)C(C(F)(F)F)=C1)C3=CC=C(C)C(/C=C/C4=CN=C(NC=C5)C5=C4OC)=C3
Molecular Weight (g/mol) 577.64
Formula Weight 577.64
Percent Purity 96.37%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.