missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HFE2 Monoclonal Antibody (OTI8F4), TrueMAB™, OriGene
Description
HFE2 Monoclonal antibody specifically detects HFE2 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | HFE2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI8F4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | Tceanc2 |
| Gene Accession No. | Q96MN5 |
| Gene Alias | 2310035L15Rik; 5230400G09Rik; AI414844; AI789733; DL M; DL-M; Fidgetin; haemojuvelin; HEMOCHROMATOSIS; hemochromatosis type 2 (juvenile); hemochromatosis type 2 (juvenile) (human homolog); hemochromatosis type 2 protein; Hemochromatosis type 2 protein homolog; hemojuvelin; Hemojuvelin BMP coreceptor; HFE 2; HFE2; HFE2A; HJV; JH; MGC23953; OTTHUMP00000059680; repulsive guidance molecule c; RGM C; RGM domain family member C; RGMC |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?