missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This antibody is a rabbit polyclonal antibody raised against full length of human HEY2.
Downstream effector of Notch signaling which may be required for cardiovascular development. Transcriptional repressor which binds preferentially to the canonical E box sequence 5′-CACGTG-3′. Represses transcription by the cardiac transcriptional activators GATA4 and GATA6.
Specifications
Specifications
| Antigen | HEY2 |
| Applications | Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | HEY2 |
| Gene Accession No. | Q9QUS4, Q9UBP5 |
| Gene Alias | bHLHb32, CHF1, GRIDLOCK, GRL, hCHF1, HERP, HERP1, HESR 2, HESR2, HEY2, hHRT2, HRT 2, HRT2, Protein gridlock homolog |
| Gene Symbols | HEY2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?