Learn More
Hexamoll (Technical Grade), TRC
Product Code. 30008078
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008078 5 g 5g
17968764 25 g 25g
17907264 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30008078 Supplier Toronto Research Chemicals Supplier No. TRCH2967105G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 166412-78-8
Molecular Formula C26 H48 O4
Molecular Weight (g/mol) 424.67
Synonym 1,2-Cyclohexanedicarboxylic acid, diisononyl ester (9CI), Bis(isononyl)cyclohexane 1,2-dicarboxylate, DINCH, Diisononyl cyclohexane-1,2-dicarboxylate, Diisononyl hexahydrophthalate, Elatur CH, Flocare 35138, Hexamoll, Hexamoll DINCH, Plastic additive 24
IUPAC Name bis(7-methyloctyl) cyclohexane-1,2-dicarboxylate
SMILES CC(C)CCCCCCOC(=O)C1CCCCC1C(=O)OCCCCCCC(C)C
Specifications
Quantity 5 g
InChI Formula InChI=1S/C26H48O4/c1-21(2)15-9-5-7-13-19-29-25(27)23-17-11-12-18-24(23)26(28)30-20-14-8-6-10-16-22(3)4/h21-24H,5-20H2,1-4H3
Formula Weight 424.36
Chemical Name or Material 1,2-Cyclohexanedicarboxylic acid, bis-isononyl ester