missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HES6 Monoclonal Antibody (OTI1H2), TrueMAB™, OriGene
Description
HES6 Monoclonal antibody specifically detects HES6 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | HES6 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI1H2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | HES6 |
| Gene Accession No. | Q96HZ4 |
| Gene Alias | AI326893; bHLHb41; bHLHc23; C-HAIRY1; class B basic helix-loop-helix protein 41; Hairy and enhancer of split 6; hes family bHLH transcription factor 6; hes6; HES-6; Transcription cofactor HES-6 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?