missing translation for 'onlineSavingsMsg'
Learn More

HES6 Monoclonal Antibody (OTI1D6), TrueMAB™, OriGene

Product Code. 17558832
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17558832 30 μL 30µL
17538832 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17558832 Supplier Origene Technologies Supplier No. TA800716S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HES6 Monoclonal antibody specifically detects HES6 in Human samples. It is validated for Western Blot

Specifications

Antigen HES6
Applications Western Blot
Classification Monoclonal
Clone OTI1D6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene KRT5
Gene Accession No. P13647
Gene Alias AI326893; bHLHb41; bHLHc23; C-HAIRY1; class B basic helix-loop-helix protein 41; Hairy and enhancer of split 6; hes family bHLH transcription factor 6; hes6; HES-6; Transcription cofactor HES-6
Host Species Mouse
Immunogen Synthetic peptide sequence is corresponding to the conserved region of human KRT5 and KRT6
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3852
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.