missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Herpes Simplex Virus UL42 Antibody (13D11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NB100-2772-0.025mg
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Herpes Simplex Virus UL42 Monoclonal antibody specifically detects Herpes Simplex Virus UL42 in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Especificaciones
| Herpes Simplex Virus UL42 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| HHV1gp061 | |
| Baculovirus-expressed Herpes Simplex Virus DNA polymerase (POL) and POL/UL42 complex. | |
| 0.025 mg | |
| Virology Bacteria and Parasites | |
| Virus | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| 13D11 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction