missing translation for 'onlineSavingsMsg'
Learn More

Heptakis(2,6-di-o-methyl)-β-cyclodextrin, MP Biomedicals™

Product Code. 11453802 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11453802

Brand: MP Biomedicals 0215732001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Heptakis(2,6-di-O-methyl)-β-cyclodextrin is a cholesterol sequestering agent

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51166-71-3
Molecular Formula C56H98O35
Molecular Weight (g/mol) 1331.365
MDL Number MFCD00011616
InChI Key QGKBSGBYSPTPKJ-UZMKXNTCSA-N
Synonym 2,6-di-o-methyl-beta-cyclodextrin, unii-5fjp73e35o, dimethyl-beta-cyclodextrin, heptakis 2,6-o-dimethyl beta-cyclodextrin, heptakis-2,6-di-o-methyl-beta-cyclodextrin, heptakis 2,6-di-o-methyl-beta-cyclodextrin, dm-beta-cyd, dimethyl, a-cyclodextrin, 2,6-dme-beta-cyclodextrin, 2,6-di-o-methyl-beta-cd
PubChem CID 10171019
SMILES COCC1C2C(C(C(O1)OC3C(OC(C(C3O)OC)OC4C(OC(C(C4O)OC)OC5C(OC(C(C5O)OC)OC6C(OC(C(C6O)OC)OC7C(OC(C(C7O)OC)OC8C(OC(O2)C(C8O)OC)COC)COC)COC)COC)COC)COC)OC)O

Specifications

Color White
Quantity 1 g
Formula Weight 1331.4
Physical Form Powder
Chemical Name or Material Heptakis(2, 6-di-o-methyl)-β-cyclodextrin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.