missing translation for 'onlineSavingsMsg'
Learn More

HEPES Hemisodium Salt, MP Biomedicals™

Product Code. 11452722 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11452722

Brand: MP Biomedicals 0215245125

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 103404-87-1
Molecular Formula C8H17N2NaO4S
Molecular Weight (g/mol) 260.28
MDL Number MFCD00036463
InChI Key RDZTWEVXRGYCFV-UHFFFAOYSA-M
Synonym hepes sodium salt, hepes hemisodium salt, sodium 2-4-2-hydroxyethyl piperazin-1-yl ethanesulfonate, unii-z9fto91o8a, 1-piperazineethanesulfonic acid, 4-2-hydroxyethyl-, monosodium salt, z9fto91o8a, sodium 4-2-hydroxyethyl piperazin-1-ylethanesulphonate, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid sodium salt, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid hemisodium salt
PubChem CID 2724248
ChEBI CHEBI:46758
IUPAC Name sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
SMILES [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1

Specifications

Color White
Assay Percent Range 1.005
Quantity 25 g
Formula Weight 249.3
Physical Form Powder
Chemical Name or Material HEPES Hemisodium Salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.