missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Hepatitis B Virus X Monoclonal Antibody (X36C)
GREENER_CHOICE

Product Code. 11578842
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11578842 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11578842 Supplier Invitrogen™ Supplier No. MA1081

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-081 detects hepatitis B x (HBx) protein from transfected human cells. MA1-081 has been successfully used in Western blot, immunoprecipitation and ELISA procedures. By Western blot, this antibody detects a single ~17 kDa protein representing HBx. This antibody is not capable of immunoprecipitating HBx complexed with HBx interacting protein (XIP). The MA1-081 immunogen is baculovirus expressed recombinant HBx.

The hepatitis B virus is an important cause of acute and chronic liver disease. It is also the most common form of parenterally transmitted viral hepatitis. The Hepatitis B X antigen is a 154 amino acid ~17 kDa multifunctional protein involved in the development of liver chirrosis and hepatocellular carcinoma. The virus is estimated to affect some 350 million people worldwide. Immunization against hepatitis B is an important component of preventative medicine and is offered to all children in approximately 30 countries. Hepatitis B virus (HBV) belongs to the Hepadnaviridae family and infection with HBV is usually more serious than infection with hepatitis A virus. In some individuals the infection is not cleared and followed by persistent intracellular viral carriage that can lead to the development of hepatocelular carcinomas. There are currently 400 million HBV carriers worldwide making HBV one of the most common human pathogens.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatitis B Virus X
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone X36C
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Alias HBV; HBVgp3; HepB; HepB X
Host Species Mouse
Immunogen Baculovirus expressed recombinant HBx.
Purification Method HPLC
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.