missing translation for 'onlineSavingsMsg'
Learn More

Scharlab Hektoen Enteric Agar

Product Code. 11471797
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11471797 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11471797 Supplier Scharlab Supplier No. 01216500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Scharlab Hektoen Enteric Agar is used for the isolation of pathogenic enterobacteria from contaminated samples.

  • ISO
  • Solid, selective and differential culture medium for isolation of pathogenic enterobacteria from contaminated samples.
  • According to ISO 21567 standard.

Especificaciones

Certifications/Compliance ISO 21567
Form Solid
Media Type Hektoen Enteric Agar
Product Type Differential Medium
Target Organism Class Gram-negative Bacteria
Formulation Meat Peptone 12.00 g/L, Yeast Extract 3.00 g/L, Bile Salts 9.00 g/L, Lactose 12.00 g/L, Sucrose 12.00 g/L, Salicin 2.00 g/L, Sodium Chloride 5.00 g/L, Sodium Thiosulfate 5.00 g/L, Ammonium Ferric Citrate 1.50 g/L, Acid Fuchsin 0.10 g/L, Bromothymol Blue 0.06 g/L, Agar 15.00 g/L
For Use With (Application) Isolation of Pathogenic Enterobacteria From Contaminated Samples
Incubation Conditions 37°C ± 1.0°C
Incubation Time 20 to 24 hr.
pH 7.5 ±0.2
Quantity 500 g
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.