Description
The thermostat can be integrated in processes by means of the Hei-PROCESS solutions using different interfaces. Furthermore, the device can be directly controlled via a Hei-VAP Expert Control or Ultimate Control rotary evaporator as recirculation chiller or can be used independently by the integrated touch display.
- 6 L tank volume for temperature stability, even under sustained load
- Precise temperature regulation and stability of ≤0.05°C
- Operating temperature range of -10°C to +70°C
- Modern 2.83 in. touch display
- Ethernet, RS 232 and RS 485 interfaces for the integration in processing solutions
Specifications
Specifications
| Capacity (Metric) | 5.5 L |
| Certifications/Compliance | CE |
| Communications | Ethernet, RS232, RS485 |
| Condenser Type | Air-cooled |
| Cooling Capacity | 600 W |
| Flow Rate | 2.9 L/min. |
| Pump Type | Centrifugal Pump |
| Temperature Range | -20°C to +70°C |
| Type | Recirculating Chiller |
| Plug Type | EU |
| Show More |