missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HEC1 Recombinant Rabbit Monoclonal Antibody (24GB415)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557102
This item is not returnable.
View return policy
Description
HEC1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Hec1 is one of several proteins involved in spindle checkpoint signaling. This surveillance mechanism assures correct segregation of chromosomes during cell division by detecting unaligned chromosomes and causing prometaphase arrest until the proper bipolar attachment of chromosomes is achieved.
Specifications
| HEC1 | |
| Recombinant Monoclonal | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O14777 | |
| NDC80 | |
| A synthesized peptide derived from human HEC1 (aa 600-642). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB415 | |
| Unconjugated | |
| NDC80 | |
| 2610020P18Rik; AV139730; HEC; HEC1; HEC-1; HEC2; HEC4; Highly expressed in cancer protein; highly expressed in cancer, rich in leucine heptad repeats; HsHec1; hsNDC80; kinetochore associated 2; kinetochore protein Hec1; Kinetochore protein NDC80 homolog; kinetochore-associated protein 2; KNTC2; NDC80; NDC80 homolog, kinetochore complex component; NDC80 homolog, kinetochore complex component (S. cerevisiae); NDC80 kinetochore complex component; NDC80 kinetochore complex component homolog; NDC80, kinetochore complex component; putative kinetochore protein NDC80 like protein; retinoblastoma-associated protein HEC; TID3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10403 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction