Learn More
Hecht Karl™ Heavy-wall Test Tubes
Product Code. 10292001
Click to view available options
Diameter (Metric):
12 mm
15.25 mm
Length (Metric):
75 mm
100 mm
160 mm
Unit Size:
Pack of 100
Pack of 200
Product Code. 10292001
Brand: Hecht Karl™ 42775024
Only null left
Add to basket
Add to basket
Manufactured with AR-Glas™ (soda glass) to minimize breakage. Karl Hecht™ Heavy-Wall Test Tubes are suitable for general laboratory use.
  • Soda lime glass
  • Round bottom
  • Rimless
  • Sized to support all laboratory needs 
Specifications
Design Type Kahn
Description Kahn test tube
Length (Metric) 75 mm
Material Soda Lime Glass
Quantity 200 Pack
Rimmed No
Diameter (Metric) 12 mm