Learn More
Hecht Karl™ Heavy-wall Test Tubes
Product Code. 10088580
Click to view available options
Diameter (Metric):
12 mm
15.25 mm
Length (Metric):
75 mm
100 mm
160 mm
Unit Size:
Pack of 100
Pack of 200
Product Code. 10088580
Brand: Hecht Karl™ 42775045
Manufactured with AR-Glas™ (soda glass) to minimize breakage. Karl Hecht™ Heavy-Wall Test Tubes are suitable for general laboratory use.
  • Soda lime glass
  • Round bottom
  • Rimless
  • Sized to support all laboratory needs 
Specifications
Design Type Round Bottom
Description Test tube
Length (Metric) 100 mm
Material Soda Lime Glass
Quantity 100 Pack
Rimmed No
Diameter (Metric) 15.25 mm