Learn More
Heidolph™ Heat-On Multi-Well Basic Package
Shop All Heidolph Instruments Products
Click to view available options
Type:
Heat-On Multi-Well Basic Package
Unit Size:
Each
Description
- Includes a multi-well holder and 3 inserts for small round bottom flasks
- The Heat-On inserts are suitable for 25 mL, 50 mL and 100 mL round bottom flasks respectively
- A max. of two flasks at a time can be received by the multi-well holder
- The max. temperature for continuous use is 260°C
- In order to measure this precisely with a temperature sensor, a small hole is provided on one side of the Heat-On Block
- Safety lifting handles are optional and are available separately
Order Info
dimProductType
Specifications
Specifications
| Type | Heat-On Multi-Well Basic Package |
| Coating | Polymer |
| Material | Aluminum |