Learn More
Masterflex™ HDPE Y Connector
Product Code. 12324598
Change view
Click to view available options
For Use With:
4.76mm
9.52mm
12.7mm
Unit Size:
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
12324598 12.7mm Pack of 10
12394588 4.76mm Pack of 10
12314598 9.52mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12324598 Supplier Masterflex™ Supplier No. 3061421
Only null left
Add to basket
Add to basket
Designed to tight tolerances, these fittings match perfectly to your tubing to ensure smooth flow and a proper fit.
  • Designed to tight tolerances
  • Made from virgin materials
  • Very good chemical resistance
  • Temp range: -53 to 65°C
  • Sterilize by ethylene oxide only
Specifications
Diameter (Metric) Inner 12.7 mm
Material HDPE
For Use With 12.7mm
Min. Operating Temperature (Metric) -53°C
Packaging Quantity 10 Pack
Max. Operating Temperature (Metric) 65°C
Product Type Y Connector