Learn More
Kautex™ HDPE Square Narrow Mouth Bottles
Product Code. 10258920
Click to view available options
Capacity (Metric):
0.1 L
0.25 L
0.5 L
1 L
2.5 L
Unit Size:
Each
Pack of 122
Pack of 247
Pack of 27
Pack of 544
Pack of 72
Product Code. 10258920
Brand: Kautex™ 2000083970
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from HDPE for strength and durability. Kautex™ HDPE Square Narrow Mouth Bottles are easily stocked and save space in the laboratory from square shape.
  • Square cross-section
  • Recessed labelling field
  • No cap. Four types of closures available separately
  • Ideal for liquids, leak-proof
Application:
  • packaging
  • sending samples
  • transport
  • storage
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 0.1 L
Color Natural
Diameter (Metric) Inner Neck 17 mm
For Use With (Application) Packaging, Sending Samples, Transport, Storage
Material HDPE
Neck Finish Narrow
Closure Included No
Mouth Narrow
Shape Square
Height (Metric) 108 mm
Product Type Narrow Neck Bottle
Diameter (Metric) 45 mm