Learn More
Kautex™ HDPE Natural Rectangular Narrow Neck Bottle
Product Code. 10258420
Change view
Click to view available options
Capacity (Metric):
0.1 L
0.25 L
0.5 L
1 L
Unit Size:
Each
Pack of 108
Pack of 205
Pack of 381
Pack of 648
Product Code. Capacity (Metric) unitSize
10258420 0.5 L Each
10571931 0.1 L Pack of 108
15901391 0.1 L Pack of 108
17878117 0.1 L Pack of 648
10248420 0.25 L Pack of 381
15911391 0.25 L Pack of 381
17838107 0.5 L Pack of 205
10282131 1 L Each
17848117 1 L Pack of 108
9 options
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Product Code. 10258420
Brand: Kautex™ 2000070522
Only null left
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
  • High stability, ribbed sides for secure grip
  • Rectangular, natural color
  • Caps are not included; sold separately
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 0.5 L
Color Natural
Diameter (Metric) Inner Neck 19 mm
For Use With (Application) Storage, Packaging, Dispensing, Mailing
Material HDPE
Neck Finish Narrow
Closure Included No
Mouth Narrow
Shape Square
Height (Metric) 180 mm
Product Type Narrow Neck Bottle