Learn More
VITLAB™ HDPE Funnel
Product Code. 10384051
Change view
Click to view available options
Diameter (Metric)Top:
400 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric)Top unitSize
10384051 400 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10384051 Supplier VITLAB™ Supplier No. 42294
Only null left
Add to basket
Add to basket
Manufactured from HDPE. VITLAB™ HDPE Funnel is suitable for filling large amounts of liquids.
  • Transparent
  • With handle 
Specifications
Diameter (Metric)Top 400 mm
Capacity (Metric) 12,500 mL
Material HDPE
Height (Metric) 365 mm
Angle 60°
Quantity 1/Pk.