Learn More
Fisherbrand™ Floating Foam Tube Racks
GREENER_CHOICE
Product Code. p-8001033 Shop All Fisherbrand Products
Click to view available options
Holds:
18 x 0.2 mL, 0.5 mL, 1.5/2 mL Tubes
24 x 1.5 to 2.0 mL Tubes
4 x 50 mL Tubes
8 x 15 mL Tubes
Color:
Blue
Green
Yellow
Shape:
Circle
Rectangular
Rectangular with Rounded Corners
Square
Unit Size:
Pack of 5
Not recommended for cleaning in autoclave.
Product Code. 11736565
Only null left
Add to basket
Add to basket
Not recommended for cleaning in autoclave.
Float tubes in water baths, tubs or beakers. Fisherbrand™ Floating Foam Tube Racks are constructed of flexible, high-density polyethylene foam for a variety of lab applications. Brightly colored, reusable racks include detachable carrying handles to easily lift samples out of baths.
  • Brightly colored foam racks are ideal for floating tubes in water baths, tubs or beakers
  • Closed cell foam resists water absorption so racks can be washed and reused
  • Include detachable carrying handles to lift samples easily out of baths
Specifications
Autoclavable Not Autoclavable
Color Blue
Type Tube Rack
For Use With (Application) For floating tubes in water baths, tubs or beakers
Holds 18 x 0.2 mL, 0.5 mL, 1.5/2 mL Tubes
Includes Detachable carrying handles
Material High-density Polyethylene Foam
No. per Pack 5 Pk.
Shape Circle
Diameter (Metric) 70 mm
Diameter (Metric) Well 9.3 mm