missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDHD2 Monoclonal Antibody (OTI3H8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502882S
This item is not returnable.
View return policy
Description
HDHD2 Monoclonal antibody specifically detects HDHD2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| HDHD2 | |
| Monoclonal | |
| 0.4 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q14765 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6775 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3H8 | |
| Unconjugated | |
| STAT4 | |
| 0610039H12Rik; 3110052N05Rik; epididymis secretory protein Li 301; haloacid dehalogenase like hydrolase domain containing 2; haloacid dehalogenase-like hydrolase domain containing 2; haloacid dehalogenase-like hydrolase domain-containing protein 2; hdhd2; HEL-S-301; hypothetical protein LOC505403; hypothetical protein LOC541346; Ier3ip1; ier3ip1 {ECO:0000250; immediate early response 3-interacting protein 1; LRRG00122; RGD1308579; UniProtKB:Q9Y5U9}; zgc:110332 | |
| Full length human recombinant protein of human STAT4 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction