missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529509
This item is not returnable.
View return policy
Description
Recommended positive controls: Jurkat, Raji, K562, NCI-H929. Predicted reactivity: Mouse (92%), Rat (91%), Chicken (81%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The biogenic amine histamine is an important modulator of numerous physiologic processes, including neurotransmission, gastric acid secretion, and smooth muscle tone. The biosynthesis of histamine from histidine is catalyzed by the enzyme L-histidine decarboxylase (HDC; EC 4.1.1.22). This homodimeric enzyme is a pyridoxal phosphate (PLP)-dependent decarboxylase and is highly specific for its histidine substrate.
Specifications
| HDC | |
| Polyclonal | |
| Unconjugated | |
| Hdc | |
| AW108189; HDC; Hdc-a; Hdc-c; Hdc-e; Hdc-s; histidine decarboxylase; L-histidine decarboxylase; MGC163399 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3067 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P19113 | |
| Hdc | |
| Recombinant protein encompassing a sequence within the center region of human HDC. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction