missing translation for 'onlineSavingsMsg'
Learn More

HDAC8-IN-1, MedChemExpress

Product Code. 16494418
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494418 10 mg 10mg
16460737 50 mg 50mg
16433076 25 mg 25mg
16427147 5 mg 5mg
16415316 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16494418 Supplier Medchem Express Supplier No. HY111342

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress HDAC8-IN-1 is a HDAC8 inhibitor with an IC50 of 27.2 nM.

HDAC8-IN-1 is a HDAC8 inhibitor with an IC50 of 27.2 nM.

Specifications

Chemical Name or Material HDAC8-IN-1
CAS 1417997-93-3
Color Light Nude
Purity Grade Notes Research
Molecular Formula C22H19NO3
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (144.76 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(NO)/C=C/C1=CC=C(OC)C=C1C2=CC=C(C3=CC=CC=C3)C=C2.[E]
Molecular Weight (g/mol) 345.39
Formula Weight 345.39
Percent Purity 99.52%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.