missing translation for 'onlineSavingsMsg'
Learn More

HDAC-IN-7, MedChemExpress

Product Code. p-200098031
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16465768

Brand: Medchem Express HY13592

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress HDAC-IN-7 (Chidamide impurity) is an impurity of Chidamide. Chidamide is a potent and orally bioavailable HDAC enzymes class I (HDAC1/2/3) and class IIb (HDAC10) inhibitor.

HDAC-IN-7 (Chidamide impurity) is an impurity of Chidamide. Chidamide is a potent and orally bioavailable HDAC enzymes class I (HDAC1/2/3) and class IIb (HDAC10) inhibitor.

Specifications

Chemical Name or Material HDAC-IN-7
Physical Form Solid
CAS 743420-02-2
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C22H19FN4O2
For Use With (Application) Cancer-programmed cell death
Synonym Chidamide impurity
SMILES O=C(NC1=CC(F)=CC=C1N)C2=CC=C(CNC(/C=C/C3=CC=CN=C3)=O)C=C2
Molecular Weight (g/mol) 390.41
Formula Weight 390.41
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.