missing translation for 'onlineSavingsMsg'
Learn More

HBX 19818, MedChemExpress

Product Code. 16459857
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16459857

Brand: Medchem Express HY17540

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress HBX 19818 is a specific inhibitor of ubiquitin-specific protease 7 (USP7), with an IC50 of 28.1 μM.

HBX 19818 is a specific inhibitor of ubiquitin-specific protease 7 (USP7), with an IC50 of 28.1 μM.

Specifications

Chemical Name or Material HBX 19818
Physical Form Solid
CAS 1426944-49-1
Color Light Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C25H28ClN3O
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 20 mg/mL (47.40 mM; Need ultrasonic)
SMILES CN(CCCNC(C1=CC=C(N=C(CCCC2)C2=C3Cl)C3=C1)=O)CC4=CC=CC=C4
Molecular Weight (g/mol) 421.96
Formula Weight 421.96
Percent Purity 98.39%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.