missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HBA1, Chicken anti-Human, Biotin, Polyclonal Antibody, Abnova™
Chicken Polyclonal Antibody
Brand: Abnova PAB29177.100ug
This item is not returnable.
View return policy
Specifications
| HBA1 | |
| Polyclonal | |
| ELISA Western Blot The optimal working dilution should be determined by the end user. | |
| hemoglobin, alpha 1 | |
| HBA1 | |
| Native purified human Hemoglobin. | |
| RUO | |
| 3039/3040/3043 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| ELISA, Western Blot | |
| Biotin | |
| In PBS, pH 7.2 (0.01% thimerosol, 20% glycerol) | |
| HBH/HBA-T3 | |
| Chicken | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction