Learn More
asecos™ Hazardous Substances Rack
Product Code. 17816376 Shop All asecos Products
Change view
Click to view available options
Capacity (Metric):
30 L
40 L
Weight (Metric):
56 kg
59 kg
64 kg
71 kg
90 kg
Unit Size:
Each
Product Code. Capacity (Metric) Weight (Metric) unitSize
17816376 30 L 56 kg Each
17506646 30 L 59 kg Each
17826506 30 L 71 kg Each
17846566 40 L 64 kg Each
17836466 40 L 90 kg Each
5 options
Product Code. 17816376
Brand: asecos™ 27853
This item requires a price update prior to being ordered.
Request a Price for this item here.
The Hazardous Substances Rack provides storage of aggressive chemicals.
  • Galvanised rack frames
  • Sturdy construction due to reinforcement cross bars
  • Storage levels in 25 mm increments, height-adjustable
  • Load capacity per storage shelf: 200 kg (evenly distributed)
  • Rack system can be extended individually
  • Flat-pack delivery with assembly instructions
  • Varioius storage level options
  • Polyethylene sumps with high resistance against aggressive chemicals (such as acids and alkalis)
Specifications
Capacity (English) 7.9 gal.
Capacity (Metric) 30 L
Material Steel
Type Drum Rack
Length (English) 41.73 in.
Length (Metric) 1060.00 mm
Width (English) 41.73 in.
Width (Metric) 1060.00 mm
Height (English) 78.74 in.
Height (Metric) 2000.00 mm
Depth (English) 25.08 in.
Depth (Metric) 637.00 mm
Weight (English) 123.46 lb.
Weight (Metric) 56 kg