Learn More
RETSCH Hard Anodized Aluminum Dividing Heads
Product Code. 10762711 Shop All Retsch Products
Click to view available options
For Use With (Equipment):
For Feed Sizes up to 10mm
For Feed Sizes up to 5mm
Includes:
10 Quick-release Sample Outlets
6 Quick-release Sample Outlets
8 Quick-release Sample Outlets
Unit Size:
Each
Product Code. 10762711
Brand: RETSCH 42.793.0009
Only null left
Specifications
Product Type Dividing Head
For Use With (Equipment) For Feed Sizes up to 5mm
Includes 8 Quick-release Sample Outlets
Material Hard Anodized Aluminum