missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Haptoglobin Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-97751
This item is not returnable.
View return policy
Description
Haptoglobin Polyclonal specifically detects Haptoglobin in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| Haptoglobin | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| binding peptide, BP, haptoglobin, haptoglobin alpha(1S)-beta, haptoglobin alpha(2FS)-beta, haptoglobin, alpha polypeptide, haptoglobin, beta polypeptide, HP2ALPHA2, HPA1S, MGC111141 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3240 | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.01 - 0.03 ug/ml, Peptide ELISA Detection limit 1:32000 | |
| NP_005134.1 | |
| HP | |
| Peptide with sequence QCKNYYKLRTEGD corresponding to internal region according to NP_005134.1, NP_001119574.1. | |
| 0.1 mg | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_005134.1; NP_001119574.1). | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction