Learn More
Hamilton™ Syringe Guide for Microliter™ and Gaslight™ Syringes
Product Code. p-183601
Click to view available options
For Use With (Equipment):
Microliter, Gastight and 700 and 1700 Series Syringes
Microliter, Gastight, 7000 Series and 25 to 500μL Syringes
Unit Size:
Each
Product Code. 10130221
Brand: Hamilton™ 14806
Designed to work with 700/7000 series MICROLITER syringes and 1700 series GASTIGHT syringes
Syringe guides are designed to prevent plunger bending and the adjustable stop prevents plunger damage.
Specifications
Product Type Syringe Guide
For Use With (Equipment) Microliter, Gastight and 700 and 1700 Series Syringes