Learn More
Hamilton™ Syringe Guide for Microliter™ and Gaslight™ Syringes
Click to view available options
For Use With (Equipment):
Microliter, Gastight and 700 and 1700 Series Syringes
Microliter, Gastight, 7000 Series and 25 to 500μL Syringes
Unit Size:
Each
Description
Designed to work with 700/7000 series MICROLITER syringes and 1700 series GASTIGHT syringes
Syringe guides are designed to prevent plunger bending and the adjustable stop prevents plunger damage.
Specifications
Specifications
| Product Type | Syringe Guide |
| For Use With (Equipment) | Microliter, Gastight, 7000 Series and 25 to 500μL Syringes |