Learn More
Hamilton™ Needle Cleaning Concentrate
Click to view available options
Volume (Metric):
70 mL
500 mL
Unit Size:
Each
Description
- Non-alkaline, non-phosphate and non-detergent based
- 70mL or 500mL bottle
- For syringes and needles
Specifications
Specifications
| Product Type | Cleaning Concentrate |
| For Use With (Equipment) | Syringe and Needle Cleaning Kit |
| Volume (Metric) | 70 mL |