Learn More
Hamilton™ Microlab™ 600 Diluter Dispenser Accessory, Screen Protector
Product Code. 15319732
Change view
Click to view available options
For Use With (Equipment):
Microlab 600 Advanced Diluter
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15319732 Microlab 600 Advanced Diluter Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15319732 Supplier Hamilton™ Supplier No. 6424101
Only null left
Add to basket
Add to basket
Designed to protect the controller screen from chemical damage if a spill occurs
  • Recommended when users are working with chemicals that could potentially harm the color touch screen user interface
  • Chemicals can easily be wiped off to limit exposure
Specifications
For Use With (Equipment) Microlab 600 Advanced Diluter