Learn More
Hamilton™ Metal Hub Blunt Point Needles (Luer Lock)
Product Code. p-4121861
Click to view available options
Needle Gauge:
14 G
17 G
18 G
19 G
20 G
21 G
22 G
22s G
23 G
24 G
25 G
26 G
26s G
33 G
Unit Size:
Pack of 6
Product Code. 11557794
Brand: Hamilton™ 91026
Can be used with TLL syringes and LT or TLL connectors.
  • Made of 304 Stainless Steel
  • Standard needle length is 51mm (2 in.)
  • Available in convenient six packs
  • Custom needle lengths and point styles are available
  • Needles are available from 10 to 33 ga
Specifications
Autoclavable Autoclavable
Material Nickel-plated Brass (Hub), Stainless Steel
Needle Gauge 26 G
Diameter (English) Inner 0.01 in.
Diameter (English) Outer 0.018 in.
Diameter (Metric) Inner 0.26 mm
Diameter (Metric) Outer 0.45 mm
Length (English) Needle 2 in.
Product Type Needle
Length (Metric) Needle 51 mm
Needle Point Style Blunt
For Use With (Equipment) PTFE Luer-lock Syringes and Luer-tip or PTFE Luer-lock Connectors