Learn More
O&M Halyard Sequential Sterilization Wrap (H300)
GREENER_CHOICE
Product Code. 30274726 Shop All Halyard Products
Change view
Click to view available options
Dimensions (L x W):
101 x 101 cm (40 x 40 in.)
114 x 114 cm (45 x 45 in.)
121 x 121 cm (48 x 48 in.)
137 x 137 cm (54 x 54 in.)
60 x 60 cm (24 x 24 in.)
76 x 76 cm (30 x 30 in.)
91 x 91 cm (36 x 36 in.)
Unit Size:
Pack of 100
Pack of 250
Pack of 300
Pack of 500
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
30274726 60 x 60 cm (24 x 24 in.) Pack of 500
30274692 76 x 76 cm (30 x 30 in.) Pack of 300
30274731 91 x 91 cm (36 x 36 in.) Pack of 300
30274752 101 x 101 cm (40 x 40 in.) Pack of 250
30274728 114 x 114 cm (45 x 45 in.) Pack of 100
30274765 121 x 121 cm (48 x 48 in.) Pack of 100
30274664 137 x 137 cm (54 x 54 in.) Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 30274726 Supplier O&M Supplier No. 10824
Halyard Sequential wraps are the industry standard for sterile processing teams.
  • Tear-, flame-, lint- and abrasion-resistant
  • POWERGUARD technology for enhanced microbial barrier protection
  • Pre-vacuum steam sterilant penetration
  • Ethylene Oxide (EO) sterilant penetration and residuals
  • Maintenance of package integrity (MPI) to meet FDA requirements
  • Instructions for Use that meet FDA guidelines
  • Class 1 flammability rating
  • Low linting
  • Made in the USA
Specifications
Autoclavable Autoclavable
Certifications/Compliance Meets NFPA Class 1
Color Blue
For Use With (Equipment) Basin sets, instrument sets, procedure trays and large linen packs (6 to 9 lb.)
Material SMS Fabric (H300)
Sterility Non-sterile
Latex Free Yes
Dimensions (L x W) 60 x 60 cm (24 x 24 in.)
Length (English) 24 in.
Width (English) 24 in.
Product Type Sterilization Wrap
Length (Metric) 60 cm
Width (Metric) 60 cm