Learn More
Gilac™ HACCP Food Contact Container
Product Code. 17968911
Click to view available options
Color:
Blue
Green
Red
White
Yellow
Unit Size:
Each
Product Code. 17968911
Brand: Gilac™ G613233
Only null left
Add to basket
Add to basket
HACCP Food Contact Container is ideal for storage, food preparation, and transporting foodstuffs.
  • HACCP round container.
  • Food contact certified.
  • For storage, food preparation and transporting foodstuffs.
  • Highly durable.
  • Reinforced bottom with anti-wear skirt.
  • Reinforced handles to prevent distortion during carrying.
  • Optional, choice of two lids.
  • External Diameter includes handles.
  • Width corresponds to diameter without handles.
Specifications
Product Type HACCP Food Container
Material Polyethylene
Color Red
Volume (English) 13.2 gal.
Volume (Metric) 50 L